Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pde-1
Entrez Gene ID
173200
Gene Name
protein-coding
Gene Aliases
CELE_T04D3.3
Chromosome Location
Chr.I: 13310887 - 13318716 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136318.5 NP_001129790.1 3 - 4 768 74
NM_060942.5 NP_493343.1 5 - 6 1073 74

View Full Product Details