Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
src-2
Entrez Gene ID
173296
Gene Name
Tyrosine protein-kinase src-2
Gene Aliases
CELE_F49B2.5
Chromosome Location
Chr.I: 14322237 - 14328965 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061101.5 NP_493502.1 3 - 4 541 71

View Full Product Details