Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nol-6
Entrez Gene ID
173520
Gene Name
Nucleolar protein 6
Gene Aliases
CELE_Y51H7C.11
Chromosome Location
Chr.II: 1425727 - 1437499 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377698.1 NP_001364780.1 15 - 16 2706 90

View Full Product Details