Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btb-9
Entrez Gene ID
185768
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_F45C12.4
Chromosome Location
Chr.II: 1728506 - 1730065 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061658.2 NP_494059.1 1 - 2 162 119

View Full Product Details