The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_062384.7 | NP_494785.1 | 4 - 5 | 897 | 98 |
GenBank mRNA | AF083231.1 | - | 4 - 5 | 835 | 98 |