Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pho-4
Entrez Gene ID
173897
Gene Name
intestinal acid PHOsphatase
Gene Aliases
CELE_T16D1.2
Chromosome Location
Chr.II: 5236251 - 5238466 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062583.7 NP_494984.5 1 - 2 114 142

View Full Product Details