Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sec-6
Entrez Gene ID
173910
Gene Name
Exocyst complex component 3
Gene Aliases
CELE_F09E5.5
Chromosome Location
Chr.II: 5359552 - 5372182 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062605.9 NP_495006.2 9 - 10 1436 125

View Full Product Details