Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aqp-2
Entrez Gene ID
174467
Gene Name
protein-coding
Gene Aliases
CELE_C01G6.1
Chromosome Location
Chr.II: 9255309 - 9261994 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381560.2 NP_001366692.1 3 - 4 694 80
NM_001383913.1 3 - 4 707 80

View Full Product Details