Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lec-2
Entrez Gene ID
174560
Gene Name
protein-coding
Gene Aliases
CELE_F52H3.7
Chromosome Location
Chr.II: 9995984 - 10017572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383927.1 6 - 7 3449 75
NM_001383928.1 6 - 7 3028 75
GenBank mRNA AB038503.1 - 3 - 4 491 75

View Full Product Details