Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl-42
Entrez Gene ID
191758
Gene Name
Ribosomal Protein, Large subunit
Gene Aliases
CELE_C09H10.1
Chromosome Location
Chr.II: 11092713 - 11094022 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063972.5 NP_496373.2 2 - 3 189 79

View Full Product Details