Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsp-1
Entrez Gene ID
174748
Gene Name
protein-coding
Gene Aliases
CELE_W02B12.3
Chromosome Location
Chr.II: 11453362 - 11455866 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373792.1 NP_001359656.1 5 - 6 783 110
NM_001381607.1 NP_001367045.1 5 - 6 786 110
NM_064041.5 NP_496442.1 4 - 5 503 110
NR_138514.1 3 - 4 1632 110

View Full Product Details