Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-29
Entrez Gene ID
174830
Gene Name
protein-coding
Gene Aliases
CELE_W03C9.4
Chromosome Location
Chr.II: 11917642 - 11935343 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064144.6 NP_496545.1 2 - 3 325 73
GenBank mRNA L39212.1 - 2 - 3 334 73

View Full Product Details