Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-29
Entrez Gene ID
174830
Gene Name
protein-coding
Gene Aliases
CELE_W03C9.4
Chromosome Location
Chr.II: 11917642 - 11935343 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129177.4 NP_001122649.1 7 - 8 1008 61
NM_001267395.3 NP_001254324.1 3 - 4 370 61
NM_064144.6 NP_496545.1 7 - 8 1023 61
GenBank mRNA L39212.1 - 7 - 8 1032 61

View Full Product Details