Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-29
Entrez Gene ID
174830
Gene Name
protein-coding
Gene Aliases
CELE_W03C9.4
Chromosome Location
Chr.II: 11917642 - 11935343 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129177.4 NP_001122649.1 9 - 10 1216 80
NM_001267395.3 NP_001254324.1 5 - 6 578 80
NM_064144.6 NP_496545.1 9 - 10 1231 80
GenBank mRNA L39212.1 - 9 - 10 1240 80

View Full Product Details