Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-37A1
Entrez Gene ID
175053
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_F01D5.9
Chromosome Location
Chr.II: 14013826 - 14016965 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064538.4 NP_496939.2 1 - 2 84 100

View Full Product Details