Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sma-4
Entrez Gene ID
175815
Gene Name
protein-coding
Gene Aliases
CELE_R12B2.1
Chromosome Location
Chr.III: 5816275 - 5820204 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047399.4 NP_001040864.1 3 - 4 464 81

View Full Product Details