Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-4
Entrez Gene ID
182259
Gene Name
protein-coding
Gene Aliases
CELE_C05D11.6
Chromosome Location
Chr.III: 6411578 - 6415656 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268009.3 NP_001254938.1 4 - 5 504 61
NM_001268010.3 NP_001254939.1 6 - 7 653 61
NM_001382895.1 6 - 7 661 61

View Full Product Details