Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pgl-2
Entrez Gene ID
182022
Gene Name
P granule abnormality protein 2
Gene Aliases
CELE_B0523.3
Chromosome Location
Chr.III: 8670543 - 8675051 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382943.1 5 - 6 1112 89
GenBank mRNA AB120730.1 - 5 - 6 1112 89

View Full Product Details