Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vha-1
Entrez Gene ID
176383
Gene Name
V-type proton ATPase 16 kDa proteolipid subunit 1
Gene Aliases
CELE_R10E11.8
Chromosome Location
Chr.III: 9781254 - 9781763 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB000917.1 - 1 - 1 641 148

View Full Product Details