Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnt-2
Entrez Gene ID
176490
Gene Name
protein-coding
Gene Aliases
CELE_Y39A1A.15
Chromosome Location
Chr.III: 10655016 - 10677545 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027665.3 NP_001022836.1 6 - 7 1971 118
NM_001027666.5 NP_001022837.1 8 - 9 1395 118
NM_001027667.5 NP_001022838.1 5 - 6 1214 118
NM_001382987.1 7 - 8 1332 118
NM_001382988.1 8 - 9 1379 118
NM_001382989.1 8 - 9 1372 118

View Full Product Details