Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnt-2
Entrez Gene ID
176490
Gene Name
protein-coding
Gene Aliases
CELE_Y39A1A.15
Chromosome Location
Chr.III: 10655016 - 10677545 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027665.3 NP_001022836.1 1 - 2 868 83

View Full Product Details