Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klp-7
Entrez Gene ID
176511
Gene Name
protein-coding
Gene Aliases
CELE_K11D9.1
Chromosome Location
Chr.III: 10805925 - 10811001 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027506.4 NP_001022677.1 1 - 2 175 109

View Full Product Details