Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-74
Entrez Gene ID
182862
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_C24H11.4
Chromosome Location
Chr.III: 11782100 - 11784110 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067124.2 NP_499525.1 5 - 6 793 64

View Full Product Details