Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aqp-11
Entrez Gene ID
191343
Gene Name
Aquaporin
Gene Aliases
CELE_ZK525.2
Chromosome Location
Chr.III: 13673280 - 13674746 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067420.6 NP_499821.2 1 - 2 190 82

View Full Product Details