Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- csn-3
- Entrez Gene ID
-
176834
- Gene Name
- COP9 signalosome complex subunit 3
- Gene Aliases
-
CELE_Y38C1AA.2
- Chromosome Location
-
Chr.IV: 194745 - 202589 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067477.8 |
NP_499878.3 |
4 - 5 |
750 |
70 |
- Gene Symbol
- Y38C1AA.12
- Entrez Gene ID
-
259547
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y38C1AA.12
- Chromosome Location
-
Chr.IV: 192256 - 202571 on Build WBcel235
View Full Product Details