Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-8
Entrez Gene ID
177494
Gene Name
protein-coding
Gene Aliases
CELE_R13A1.4
Chromosome Location
Chr.IV: 7197153 - 7203660 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307363.3 NP_001294292.1 15 - 16 1656 110
NM_001307364.3 NP_001294293.1 14 - 15 1662 110
NM_001307365.3 NP_001294294.1 13 - 14 1539 110
NM_068737.2 NP_501138.1 15 - 16 1659 110
GenBank mRNA U76402.1 - 15 - 16 1659 110

View Full Product Details