Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtr-4
Entrez Gene ID
177824
Gene Name
mRNA transport homolog 4
Gene Aliases
CELE_W08D2.7
Chromosome Location
Chr.IV: 9825694 - 9830620 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069356.6 NP_501757.1 3 - 4 397 101

View Full Product Details