Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-12
Entrez Gene ID
188050
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_T04B2.4
Chromosome Location
Chr.IV: 10051286 - 10053650 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069419.2 NP_501820.2 5 - 6 712 158

View Full Product Details