Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- srb-17
- Entrez Gene ID
-
184054
- Gene Name
- Serpentine receptor class beta-17
- Gene Aliases
-
CELE_F01D4.7
- Chromosome Location
-
Chr.IV: 10454318 - 10457237 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069529.5 |
NP_501930.4 |
3 - 4 |
242 |
168 |
- Gene Symbol
- F01D4.8
- Entrez Gene ID
-
184055
- Gene Name
- PALP domain-containing protein
- Gene Aliases
-
CELE_F01D4.8
- Chromosome Location
-
Chr.IV: 10450730 - 10456353 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069528.3 |
NP_501929.1 |
1 - 2 |
128 |
168 |
View Full Product Details