Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fmo-1
Entrez Gene ID
177954
Gene Name
Dimethylaniline monooxygenase [N-oxide-forming]
Gene Aliases
CELE_K08C7.2
Chromosome Location
Chr.IV: 10661857 - 10664404 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069567.7 NP_501968.2 7 - 8 1632 91
GenBank mRNA AJ582070.1 - 6 - 7 1521 91

View Full Product Details