Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-18
Entrez Gene ID
176992
Gene Name
protein-coding
Gene Aliases
CELE_F55A8.1
Chromosome Location
Chr.IV: 1913446 - 1917841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307519.4 NP_001294448.1 1 - 2 155 70
NM_067742.7 NP_500143.1 1 - 2 155 70
GenBank mRNA AF353302.1 - 1 - 2 129 70

View Full Product Details