Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-51
Entrez Gene ID
189146
Gene Name
protein-coding
Gene Aliases
CELE_W03B1.7
Chromosome Location
Chr.IV: 4336823 - 4341306 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307374.3 NP_001294303.1 5 - 6 808 72
NM_001383076.1 5 - 6 822 72
NM_068143.5 NP_500544.3 4 - 5 854 72

View Full Product Details