Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wapl-1
Entrez Gene ID
177211
Gene Name
protein-coding
Gene Aliases
CELE_R08C7.10
Chromosome Location
Chr.IV: 4442362 - 4445740 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372798.2 NP_001360111.1 5 - 5 767 71
NM_001380190.1 NP_001368333.1 5 - 6 773 71
NM_068165.6 NP_500566.1 5 - 6 773 71
NM_068166.8 NP_500567.1 5 - 6 767 71

View Full Product Details