Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lam-1
Entrez Gene ID
177292
Gene Name
protein-coding
Gene Aliases
CELE_W03F8.5
Chromosome Location
Chr.IV: 5173332 - 5183746 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068333.6 NP_500734.3 6 - 7 2865 102

View Full Product Details