Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wht-5
Entrez Gene ID
178182
Gene Name
ABC transporter domain-containing protein
Gene Aliases
CELE_F19B6.4
Chromosome Location
Chr.IV: 12337577 - 12340458 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069951.5 NP_502352.1 2 - 3 222 71

View Full Product Details