Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-3
Entrez Gene ID
178252
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_F49E11.11
Chromosome Location
Chr.IV: 13059584 - 13060695 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070103.5 NP_502504.1 2 - 3 219 66

View Full Product Details