Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- srw-63
- Entrez Gene ID
-
189604
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y37A1B.10
- Chromosome Location
-
Chr.IV: 14014024 - 14015944 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070285.2 |
NP_502686.2 |
1 - 2 |
737 |
98 |
View Full Product Details