Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- srv-16
- Entrez Gene ID
-
190892
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y105C5B.10
- Chromosome Location
-
Chr.IV: 15931822 - 15933914 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070498.2 |
NP_502899.2 |
1 - 2 |
227 |
102 |
View Full Product Details