Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-29
Entrez Gene ID
178581
Gene Name
UDPGT domain-containing protein
Gene Aliases
CELE_Y49C4A.8
Chromosome Location
Chr.V: 696523 - 699589 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383270.1 6 - 7 1833 109

View Full Product Details