Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-13
Entrez Gene ID
191968
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C24B9.8
Chromosome Location
Chr.V: 2725902 - 2727201 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071339.1 NP_503740.1 4 - 5 769 130

View Full Product Details