Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srt-35
Entrez Gene ID
185925
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_F47D2.4
Chromosome Location
Chr.V: 4261485 - 4264616 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071775.1 NP_504176.1 2 - 3 150 140

View Full Product Details