Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grl-13
Entrez Gene ID
185200
Gene Name
protein-coding
Gene Aliases
CELE_F32D1.4
Chromosome Location
Chr.V: 4362668 - 4364650 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330858.4 NP_001317808.1 2 - 2 336 105
NM_071800.3 NP_504201.2 2 - 3 369 105

View Full Product Details