Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ncx-2
Entrez Gene ID
178918
Gene Name
protein-coding
Gene Aliases
CELE_C10G8.5
Chromosome Location
Chr.V: 5303689 - 5311270 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028472.5 NP_001023643.1 1 - 2 121 69
NM_001028473.5 NP_001023644.1 1 - 2 130 69
NM_072013.8 NP_504414.4 1 - 2 134 69
NM_072014.7 NP_504415.2 1 - 2 134 69

View Full Product Details