Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mcp-1
Entrez Gene ID
178982
Gene Name
protein-coding
Gene Aliases
CELE_C10F3.4
Chromosome Location
Chr.V: 5961807 - 5969175 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028468.4 NP_001023639.1 5 - 6 505 112
NM_001380650.1 NP_001367490.1 5 - 6 572 112

View Full Product Details