Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-16
Entrez Gene ID
178983
Gene Name
3 beta-hydroxysteroid dehydrogenase dhs-16
Gene Aliases
CELE_C10F3.2
Chromosome Location
Chr.V: 5983745 - 5985542 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072153.7 NP_504554.1 2 - 3 772 100

View Full Product Details