Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttn-1
Entrez Gene ID
266969
Gene Name
protein-coding
Gene Aliases
CELE_W06H8.8
Chromosome Location
Chr.V: 6120909 - 6202632 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029029.5 NP_001024200.1 13 - 14 9364 74
NM_001359777.3 NP_001346724.1 12 - 13 8074 74

View Full Product Details