Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-89
Entrez Gene ID
191878
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_F25E5.14
Chromosome Location
Chr.V: 7463453 - 7464796 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072518.2 NP_504919.2 1 - 2 221 108

View Full Product Details