Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-22
Entrez Gene ID
179485
Gene Name
protein-coding
Gene Aliases
CELE_F29F11.5
Chromosome Location
Chr.V: 10671918 - 10675769 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083273.4 NP_001076742.1 4 - 5 1193 149
NM_001083274.3 NP_001076743.1 1 - 2 84 149
NM_001083275.4 NP_001076744.1 3 - 4 288 149
NM_001356738.3 NP_001343841.1 5 - 6 576 149
NM_001356739.4 NP_001343842.1 3 - 3 298 149

View Full Product Details