Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-33
Entrez Gene ID
179538
Gene Name
protein-coding
Gene Aliases
CELE_H19N07.2
Chromosome Location
Chr.V: 11113907 - 11118824 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028841.5 NP_001024012.1 5 - 6 2233 75
NM_001269365.3 NP_001256294.1 5 - 6 2232 75
NM_001269366.3 NP_001256295.1 5 - 6 2232 75
NM_001269367.3 NP_001256296.1 5 - 6 2233 75
NM_001269368.3 NP_001256297.1 5 - 6 2233 75
NM_001269369.3 NP_001256298.1 5 - 6 2228 75
NM_001307753.4 NP_001294682.1 5 - 6 2242 75
NM_001307754.4 NP_001294683.1 5 - 6 2241 75
NM_001307755.3 NP_001294684.1 5 - 6 2241 75
NM_001307756.3 NP_001294685.1 5 - 6 2235 75
NM_001307757.3 NP_001294686.1 5 - 6 2241 75
NM_001307758.3 NP_001294687.1 5 - 6 2241 75
NM_001307759.4 NP_001294688.1 5 - 6 2216 75
NM_001307760.4 NP_001294689.1 5 - 6 2237 75
NM_073424.4 NP_505825.1 5 - 6 2226 75
NM_073425.8 NP_505826.1 5 - 6 2232 75

View Full Product Details