Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-36
Entrez Gene ID
3565335
Gene Name
Ligand-Gated ion Channel
Gene Aliases
CELE_F07B10.5
Chromosome Location
Chr.V: 11256521 - 11258996 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028595.2 NP_001023766.1 4 - 5 579 138

View Full Product Details